Learn More
Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps
Autoclavable polypropylene
Specifications
Autoclavable Yes
Closure Material Polypropylene
Closure Type Screw
Color Natural
Material Polypropylene
View More Specs
Description
Description
Autoclavable polypropylene
  • Withstand a maximum force of 20,000 × G
  • Free-standing with cap
  • Natural color
Order Info
500 per pack, 10 packs per case
Specifications
Specifications
Yes
Screw
Polypropylene
Polypropylene
Natural
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps