Learn More
Fisherbrand™ Gel-Loading Aerosol-Barrier Tips
GREENER_CHOICE
Help minimize liquid retention
Brand:  Fisherbrand™ F010R204S100FIS
53.36 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 11967734
  • £70.25 / Pack of 1020
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • No RNase or DNase contamination
  • Natural color
  • For use with, but not limited to: Biohit Proline™ Electronic, BrandTech™ Transferpette™, Eppendorf™ Reference 2000, Eppendorf Research 2100, Thermo Finnpipette™ Focus, Fisherbrand™ Finnpipette™, Fisherbrand Finnpipette II, Genemate™ ISC Bio Express, Hamilton SoftGrip™, Jencons Sealpette™ Electronic, Labnet Labpette™ Digital, Mettler Toledo VoluMate™, Nichiryo 5000, Oxford™ Benchmate™, Pipetman™, Rainin™, Vistalab Ovation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 to 100 μL
Sterile
2.7 in.
Help minimize liquid retention
Gel Loading
Polypropylene
Filtered
Natural
69 mm
Rack
Racked