missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ General Purpose Liquid-in-Glass Partial Immersion Thermometers
Brand: H-B Instrument™ B60760-1300
This item is not returnable.
View return policy
Specifications
ISO/IEC 17025:2005, A2LA, NIST | |
Partial Immersion | |
305 mm | |
Blue Organic Liquid | |
-50°C | |
2°C | |
76 mm |
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries | |
12 in. | |
Glass | |
50°C | |
-50°C to +50°C | |
2° |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- H-B DURAC PLUS ULTRA LOW LIQUID-IN-GLASS LABORATOR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : acetone