missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Heavy-Duty/Utility Funnels

Description
Ideal for continuous lab work and heavy-duty jobs
- Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
- Autoclavable to 120°C (248°F)

Specifications
15.5cm | |
Polypropylene | |
Autoclavable |
410 mL | |
17.7 cm |
Product Suggestions
Customers who viewed this item also viewed
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Heavy-Duty/Utility Funnels > Top dia. x H: 155 x 177mm; Capacity: 410mL
Spot an opportunity for improvement?Share a Content Correction