missing translation for 'onlineSavingsMsg'
Learn More

Humidity Sponge™ Dessicant Window Type, Fisherbrand™

Meet all drying requirements with these humidity sponges. Fisherbrand™ Dessicant Window Type Humidity Sponges™ are non-toxic and Class M3.5 cleanroom-compatible.

Brand:  Fisherbrand

Code : 50

Additional Details : Weight : 0.40860kg

Product Code. 11724366

  • £98.95 / Pack of 40
In Stock
to see stock.

Description

Description

Meet all drying requirements with these humidity sponges. Fisherbrand™ Dessicant Window Type Humidity Sponges™ are non-toxic and Class M3.5 cleanroom-compatible.

  • Bags result in 37% moisture absorption
  • Indicators show when moisture capacity has been reached
  • One bag will retain 16% moisture in a container with a volume of two gallons (476 cu. in.)
  • Bag size: 7.6 x 7.6 x 0.6 cm (3 x 3 x 1/4 in.) 
Specifications

Specifications

Fisherbrand™ Humidity Sponge™ Dessicant
Sodium Calcium Aluminosilicate Hydrate
Each bag weighs 10 g (0.35 oz.)
1302-78-9
40 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers