Learn More
Humidity Sponge™, Fisherbrand™
Humidity Sponge Dessicant; Fisherbrand; Reusable Type
Brand: Fisherbrand 07193-13
This item is not returnable.
View return policy
Description
Humidity Sponge Dessicant; Fisherbrand; Reusable Type
- Non-toxic desiccant
- Reusable
- Material is made of DuPont's Tyvek™ breathable, high-density polyethylene
- Perfect for drugs, food, electronics, optics and equipment
- Bags are guaranteed to retain 16% moisture by weight, typical results are 37% moisture absorption
- Desiccant meets US FDA requirements
- Certified ISO Class 5 (Class M3.5) cleanroom compatible
Specifications
Fisherbrand™ Humidity Sponge™ Dessicant | |
HDPE | |
Certified |
1302-78-9 | |
40 Pk. | |
11 g |