missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hygrometer Replacement Battery

Hygrometer Replacement Battery for use with Hygrometers.

Brand:  Fisherbrand™ 90023-39

18.22 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15338754

  • £26.40 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

Hygrometer Replacement Battery for use with Hygrometers.

  • Come with lithium, button cell and “AAA” alkaline battery
  • The electrical requirements are 3 V, 1.5 V and 1.5 V respectively
Especificaciones

Especificaciones

Button Cell Battery
Button
-58°F to +302°F
1.5 V
357
For hygrometers
-50°C to +150°C
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales