missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isoelectric Focusing Unit
Complete IEF Unit with tank, cooling plate, electrodes.
Brand: Fisherbrand™ IEF-SYS
Includes: Two removable gel trays, 15 sample combs, aluminum casting gates, coloured loading strips
Description
- Operates in one of three modes: wick based electrophoresis; pre-cast horizontal gels, isoelectric focusing
- Unit dimensions (D x W x H): 41 x 46 x 11.5cm
- Inner tank dimensions (D X W X H): 31 x 37 x 8.5cm
- Electrode frame ( L x W x H): 27 x 27 x 1cm
- Temperature for cooling plate during IEF with IEF gels: 4°C
- Robust acrylic construction
- Safety lid with integral power leads
- Shrouded power connectors
- Long life 99.99% pure replaceable platinum electrodes
- Gold plate terminal with stainless steel washers to resist corrosion
- Printed grid - permits migration rates to be measured
- Integral cooling coil - allows for either recirculating chiller or for water connection to chill ceramic plate to desired temperature
- Large cooled ceramic top plate - provides uniform cooling of gel platform
- Two sets of electrodes
- Tank mounted - for running 'wick' gels using the lower buffer chambers
- Detachable
- Hollow frame - for IEF where strips are placed directly onto ceramic plate
- Glass bottomed frame - for pre-cast horizontal gels
- Buffer drainage ports - inbuilt shut-off valve is opened when drainage tube is connected to empty buffer chamber
Specifications
IEF Unit | |
30 x IPG strips, 1 x 245 x 125 mm, 2 x 125 x 125 mm IEF gels |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction