Learn More
Fisherbrand™ Infecon™ 2000 and 2050 Infectious/Diagnostic Shippers
For shipping test tubes containing infectious substances or diagnostic specimens
Brand:  Fisherbrand™ 19075384B
 View more versions of this product
Product Code. 15502066
  • £133.00 / Pack of 6
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For shipping test tubes containing infectious substances or diagnostic specimens
  • Bubble-wrap compartment
  • Liquid-tight, plastic secondary container withstands pressure up to 95kPa
  • U.N. markings and a label for both infectious substances and diagnostic specimens
Compliance
Complies with ICAO/IATA Packing Instruction 602
Specifications
Specifications
Complies with ICAO/IATA Packing Instruction 602
For shipping test tubes containing infectious substances or diagnostic specimens
4 x 4 x 8 in.
Infecon 2000, Infectious/Diagnostic Shipper