missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Instant Sealing Sterilization Pouches

No tape or heat sealing needed

Brand:  Fisherbrand™

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 18548066

  • £7.87 / Pack of 200
Estimated Shipment: 03-09-2024
to see stock.

Description

Description

No tape or heat sealing needed

Integral sealing strip eliminates the need to tape or heat seal pouches. Simply peel off protective strip, fold along perforation and press firmly to seal pouch.

  • Triple heat-processed seams provide excellent tear and puncture resistance
  • Suitable for steam or EtO gas sterilization
  • Indicator arrows change color when sterilization cycle is complete
  • Made of paper/plastic; medical-grade paper side guards against puncture
  • 2mil-thick blue transparent polyester/polypropylene laminate film side allows contents to be identified quickly and shows any compromise in the film
  • Corner tack seals prevent contamination in the seal area
  • Thumb notch makes opening pouch easy

Specifications

Specifications

Blue
Non-sterile
3.5 in.
5 in.
Paper, Plastic
2 mil
9 cm
13.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers