missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ L-Shaped Cell Spreaders

Product Code. 15625467 Shop All Fisherbrand Products
Click to view available options
Quantity:
1/Pouch, 100 Pouches/Cs.
10/Pouch, 50 Pouches/Cs.
missing translation for 'unitSize'
Pack of 100
Pack of 500

Product Code. 15625467

Only null left
Add to basket
Edge
Add to basket

Ideal for even spreading of bacterial culture

For spreading of bacterial culture on agar plates

  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
  • L-shape allows application on the entire Petri dish or plate surface
  • Presterilized blue polypropylene eliminates the need for flaming or autoclaving

Specifications

Disposable Single-use
Color Blue
For Use With (Equipment) Agar Culture Plate
Packaging Individually Packaged
Quantity 1/Pouch, 100 Pouches/Cs.
Shape L-Shape
Material Polypropylene
Product Type Cell Spreader
Sterility Sterile
For Use With (Application) Ideal for even spreading of bacterial cultures, smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.