missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ L-Shaped Cell Spreaders

Ideal for even spreading of bacterial culture

£16.20 - £69.70

Specifications

Color Blue
Material Polypropylene
Product Type Cell Spreader
Sterility Sterile
For Use With (Equipment) Agar Culture Plate
View More Specs

Products 2
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
15615467
View Documents
Fisherbrand™
14665230
Cell Spreader 10 each per pouch, 50 pouches/case
£69.70
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
15625467
View Documents
Fisherbrand™
14665231
Cell Spreader 1 each per pouch, 100 pouches/case
£16.20
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

For spreading of bacterial culture on agar plates

  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
  • L-shape allows application on the entire Petri dish or plate surface
  • Presterilized blue polypropylene eliminates the need for flaming or autoclaving

Specifications

Specifications

Blue
Cell Spreader
Agar Culture Plate
Single-use
Polypropylene
Sterile
L-Shape
Ideal for even spreading of bacterial cultures, smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.