missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Lab Hazard Warning Label Refills
Printed in safety colors.
Brand: Fisherbrand™ 15-933
This item is not returnable.
View return policy
Description
Printed in safety colors.
For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).
- Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
- Serrated tear-off edge for easy tape separation
- Approximately 200 imprints per roll
Order Info
Each tape roll measures 0.50 in. W x 22 yd. L (13mm x 20m)
Specifications
Assorted | |
Corrosive Warning Refill Tape | |
Printed in safety colors | |
57 yd. | |
0.5 in. |
Tape | |
Corrosive | |
4/Cs. | |
20.3 m |