Learn More
Fisherbrand™ Lid for 96/384 Well Polystyrene Plates
GREENER_CHOICE
Protect samples from contamination and evaporation during assay processing, incubation or storage
Brand:  Fisherbrand™ 236400
140.02 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 11927964
  • £190.65 / Pack of 180
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage
  • Clear
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, array and storage
96-well Plate
Polystyrene
180
Plate Lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Lid for 96/384 Well Polystyrene Plates > Lid; Nonsterile; Clear