Learn More
Fisherbrand™ Disposable Anti-Static Shoe Covers
Shop All Fisherbrand Products
Click to view available options
:
Blue
Description
Conductive carbon strip helps eliminate static buildup
- Single-use
- Elastic ankle
- 42 gms polypropylene
- With carbon dissipative strip sewn in
- Latex-free
- Blue
Specifications
Specifications
| Size | Universal |
| Material | Polypropylene |
| Latex Free | Yes |
| Color | Blue |
| Disposable | Single-use |
| Closure Type | Elastic Ankle |
| Sole Type | Anti-static |