missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Micro Slide Storage Cabinet Bases

Bases for Fisherbrand Micro Slide Storage Cabinets

Brand:  Fisherbrand™ 07212104

 View more versions of this product

Product Code. 16237260

  • £264.00 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Bases for Fisherbrand Micro Slide Storage Cabinets

For use with Fisherbrand Micro Slide Storage Cabinets (Cat. No. 07-212-100 through -103)

  • Optional bases in models with legs or without legs
  • Available in dark beige, tan, green, or white
  • 47.6L x 40W x 12.1cm H (18.75 x 15.75 x 4.75 in.)
  • Specifications

    Specifications

    Beige
    476.25 mm
    Fisherbrand Micro Slide Storage Cabinets
    120.65 mm
    Metal
    400.05 mm
    18.75 in.
    Microscope slide
    4.75 in.
    Base and legs
    15.75 in.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.