missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microscope Slide Boxes with Numbered Slots

Product Code. 11751476 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Red
White
Slide Capacity:
25
Unit Size:
Each

Product Code. 11751476

Only null left
Add to basket
Edge
Add to basket

Store and transport 25 or 100 slides safely with durable hinged cases. Fisherbrand™ Microscope Slide Boxes with Numbered Slots keeps standard microscope slides in secure slots that correspond to the numbered identification card. Stackable boxes close securely with latch lock.

Store and transport 25 or 100 slides safely with durable hinged cases. Fisherbrand™ Microscope Slide Boxes with Numbered Slots keeps standard microscope slides in secure slots that correspond to the numbered identification card. Stackable boxes clos

  • Constructed of durable polypropylene or paperboard, with no lining
  • Hinged lids are held securely closed by latch lock
  • Stacking feet provide maximize stability
  • Numbered slide identification card included for easy identification

Specifications

Type Slide Holder
Slide Capacity 25
Closure Type Hinged lid with latch lock
Color Red
Device Holds 25
Dimensions (L x W x H) 141 x 92 x 30 mm
For Use With (Equipment) Standard Microscope Slides (25 x 75 mm)
Includes Numbered index card
Material Polypropylene
Stackable Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.