Learn More
Fisherbrand™ Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
£25.00 - £320.00
Product Code | Brand | Capacity (Metric) | Closure Material | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Material | Shape | Price | Quantity & Availability | |||
12029977
|
Fisherbrand™
|
118 mL | Polypropylene | Short, Wide |
£320.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11709388
|
Fisherbrand™
|
148 mL | Polypropylene | Tall, Thin |
£165.00
Pack of 300 |
Estimated Shipment: 13-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12049977
|
Fisherbrand™
|
237 mL | LDPE | Short, Wide |
£68.05
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11719388
|
Fisherbrand™
|
237 mL | LDPE | Short, Wide |
£32.85
Pack of 100 |
Estimated Shipment: 13-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11974375
|
Fisherbrand™
|
237 mL | LDPE | Tall, Thin |
£44.35
Pack of 100 |
Estimated Shipment: 24-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12029957
|
Fisherbrand™
|
237 mL | LDPE | Tall, Thin |
£47.55
Pack of 100 |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12089977
|
Fisherbrand™
|
473 mL | LDPE | Short, Wide |
£52.70
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11994375
|
Fisherbrand™
|
473 mL | LDPE | Tall, Thin |
£55.57
Pack of 100 |
Estimated Shipment: 24-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12089947
|
Fisherbrand™
|
946 mL | LDPE | Tall, Thin |
£104.00
Pack of 100 |
Estimated Shipment: 16-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12089957
|
Fisherbrand™
|
1100 mL | LDPE | Short, Wide |
£81.90
Pack of 100 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
12009957
|
Fisherbrand™
|
1900 mL | LDPE | Tall, Thin |
£66.00
Pack of 50 |
Estimated Shipment: 22-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12009967
|
Fisherbrand™
|
1900 mL | LDPE | Tall, Thin |
£82.00
Pack of 50 |
Estimated Shipment: 13-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12039957
|
Fisherbrand™
|
2500 mL | LDPE | Tall, Thin |
£53.40
Pack of 25 |
In Stock
Log in to see stock. |
|||
12069967
|
Fisherbrand™
|
2500 mL | LDPE | Tall, Thin |
£41.70
Pack of 25 |
Estimated Shipment: 24-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12099957
|
Fisherbrand™
|
5100 mL | LDPE | Short, Wide |
£25.00
Pack of 10 |
In Stock
Log in to see stock. |
|||
12089967
|
Fisherbrand™
|
5100 mL | LDPE | Tall, Thin |
£35.05
Pack of 10 |
Estimated Shipment: 24-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Descrizione
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
- Resist freezing and boiling
- Stackable for compact storage
- Select containers are autoclavable (see ordering table); lids are not autoclavable
- Short/wide style
- Tall/thin style
- Graduated
Specifica
Specimen Container |