Learn More
Fisherbrand™ Nonsterile Cotton Balls
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Brand:  Fisherbrand™ 22456883
 View more versions of this product
Product Code. 15810000
  • £141.00 / Pack of 2000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
  • Uniform in size and shape
  • Made of pure, natural, bleached cotton for increased absorbency
  • Available in sizes medium, large, extra large and bulk roll
Order Info
For the sterile version of 22-456-880, please order 22-456-885.
Specifications
Specifications
Cotton Balls
Large
3.75 lb.
Non-sterile
2000 Cs.
0.004 kg