missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
GREENER_CHOICE

Choose skirted- or conical-bottom style.

£15.85 - £15.90

Specifications

Sterility Non-sterile
Closure Type Screw Cap
View More Specs

Products 2
Product Code Brand Color Capacity (Metric) Max. RCF Bottom Shape Price Quantity & Availability  
Product Code Brand Color Capacity (Metric) Max. RCF Bottom Shape Price Quantity & Availability  
15321129
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
516-FIS
Natural 0.5 mL 30,000 x g Skirted
£15.90
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
15331129
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
528-FIS
Natural 1.5 mL 18,000 x g Skirted
£15.85
Pack of 500
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
Description

Description

Choose skirted- or conical-bottom style.

  • Threaded to accept colored screw caps (sold separately)
  • Siliconized/low-retention conical-bottom tubes also available
Note: For caps, see Fisherbrand™ Colored Screw Caps. 
Specifications

Specifications

Non-sterile
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.