missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator

Printed in English and Spanish.

Brand:  Fisherbrand™ 01814C

 View more versions of this product

Product Code. 18326513

  • £245.00 / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Printed in English and Spanish

 

  • Maximum temperature of 140°C (285°F); use at 125°C (257°F)
  • Come with wire-tie closures
  • With sterilization indicator

 

Specifications

Specifications

ASTM D1709, Dart impact
Orange
89 cm
Autoclavable
35 in.
Biohazard Waste Bags with indicator
Wire Tie
35 x 25 in. (89 x 63.5 cm)
63.5 cm
25 in.
Polypropylene
140°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers