missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator
Printed in English and Spanish.
Brand: Fisherbrand™ 01814A
This item is not returnable.
View return policy
Description
Printed in English and Spanish
- Maximum temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
- With sterilization indicator
Specifications
ASTM D1709, Dart impact | |
Orange | |
48 cm | |
Autoclavable | |
19 in. | |
Biohazard Waste Bags with indicator |
Wire Tie | |
19 x 14 in. (48 x 36 cm) | |
36 cm | |
14 in. | |
Polypropylene | |
140°C |