missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator

Printed in English and Spanish.

Brand:  Fisherbrand™ 018151

 View more versions of this product

Product Code. 18538066

  • £15.15 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Printed in English and Spanish

 

  • Maximum temperature of 140°C (285°F); use at 125°C (257°F)
  • Come with wire-tie closures
  • With sterilization indicator

 

Specifications

Specifications

ASTM D1709, Dart impact
Orange
28 cm
Autoclavable
11 in.
Biohazard Waste Bags with indicator
Wire Tie
11 x 8.5 in. (28 x 21.5 cm)
21.5 cm
8.5 in.
Polypropylene
140°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator > Size: 8.5W x 11 in. L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.