missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Test Paper Rolls
General purpose pH papers in efficient 15 ft. rolls
£51.95
Specifications
For Use With (Application) | Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications |
---|---|
Storage Requirements | Room Temperature |
Type | pH Test Paper |
Product Code | Brand | pH Range | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Quantity | Price | Quantity & Availability | ||||
17785738
|
Fisherbrand™
|
0 to 14 | 100 Tests/Roll, 700 Total |
£51.95
Pack of 700 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
General purpose pH papers in efficient 15 ft. rolls
- Convenient dispenser allows you to select the right amount of pH paper for each application
- Easy-to-use—Dip paper in solution to be tested and match to color chart immediately
- Highly accurate and well suited for most applications
- Each SKU is packed in box of five clear plastic dispensers
- Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
Order Info
15 ft. Roll provides about 100 tests.
Specifications
Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications | |
Room Temperature | |
pH Test Paper |