Learn More
Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Product Code | Brand | Color | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | For Use With (Equipment) | Price | Quantity & Availability | ||||
17555156
|
Fisherbrand™
14-955-256 |
Red | Standard taper Glass joint size 29 |
N/A
|
N/A | ||||
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
10/Pk. | |
Joint Clip | |
Plastic |