missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Brand: Fisherbrand™ 01-814-3
This item is not returnable.
View return policy
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
ASTM D1709-85 | |
This bag has no biohazard symbol, and no indicator. | |
24 x 36 in. (61 x 91 cm) | |
61 cm | |
24 in. | |
1.50 mil | |
Autoclavable Bags |
None | |
Natural | |
91 cm | |
Autoclavable | |
36 in. | |
Polyethylene | |
127°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction