missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Brand: Fisherbrand™
Additional Details : Weight : 0.01100kg
Description
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
OSHA, 29 CFR1910.1030 | |
Red with Black Print | |
91 cm | |
Autoclavable | |
24 in. | |
4 mil | |
Autoclavable Bags |
Elastic | |
36 x 24 in. (91 x 61 cm) | |
61 cm | |
Printed in English and Spanish | |
36 in. | |
Polyethylene | |
127°C |