missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Brand: Fisherbrand™ 01827A
This item is not returnable.
View return policy
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
OSHA, 29 CFR1910.1030 | |
Clear with Red Print | |
30 cm | |
Autoclavable | |
8 in. | |
1.50 mil | |
Autoclavable Bags |
Elastic | |
12 x 8 in. (30 x 20 cm) | |
20 cm | |
Printed in English and Spanish | |
12 in. | |
Polyethylene | |
127°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction