missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Brand:  Fisherbrand™ 01827C

 View more versions of this product

Product Code. 18366513

  • £181.00 / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications

Specifications

OSHA, 29 CFR1910.1030
Clear with Red Print
76 cm
Autoclavable
24 in.
1.50 mil
Autoclavable Bags
Elastic
30 x 24 in. (76 x 61 cm)
61 cm
Printed in English and Spanish
30 in.
Polyethylene
127°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.