missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand: Fisherbrand™ 22044560
This item is not returnable.
View return policy
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Twist Tie | |
36 x 24 in. (91 x 61 cm) | |
61 cm | |
Biohazard symbol and text in English and Spanish | |
36 in. | |
Polyethylene |
Clear with Red Print | |
91 cm | |
Autoclavable | |
24 in. | |
1.50 mil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction