missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
£41.37 - £180.22
Specifications
Closure Type | Twist Tie |
---|---|
Autoclavable | Autoclavable |
Includes | Biohazard symbol and text in English and Spanish |
Material | Polyethylene |
Product Code | Brand | Color | Dimensions (L x W) | Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (L x W) | Thickness | Price | Quantity & Availability | |||
18356513
|
Fisherbrand™
|
Clear with Red Print | 30 x 24 in. (76 x 61 cm) | 1.50 mil |
£88.33
Pack of 100 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18568056
|
Fisherbrand™
|
Clear with Red Print | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
£41.37
Pack of 100 |
Estimated Shipment: 06-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18588056
|
Fisherbrand™
|
Red with Black Print | 12 x 8 in. (30 x 20 cm) | 1.25 mil |
£127.43
Pack of 1000 |
Estimated Shipment: 06-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18356533
|
Fisherbrand™
|
Red with Black Print | 19 x 14 in. (48 x 36 cm) | 1.25 mil |
£60.17
Pack of 200 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18376533
|
Fisherbrand™
|
Clear with Red Print | 36 x 24 in. (91 x 61 cm) | 1.50 mil |
£104.88
Pack of 100 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18336513
|
Fisherbrand™
|
Red with Black Print | 23 x 19 in. (58 x 48 cm) | 1.25 mil |
£91.43
Pack of 200 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18316543
|
Fisherbrand™
|
Red with Black Print | 35 x 25 in. (89 x 64 cm) | 1.50 mil |
£180.22
Pack of 200 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18396513
|
Fisherbrand™
|
Red with Black Print | 36 x 45 in. (91 x 114 cm) | 1.50 mil |
£162.34
Pack of 100 |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Twist Tie | |
Autoclavable | |
Biohazard symbol and text in English and Spanish | |
Polyethylene |