missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Waste Bags: Plain

Plain style (without print).

Brand:  Fisherbrand™ 018266

 View more versions of this product

Product Code. 16418115

  • £130.00 / Pack of 200
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Plain style bags

  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures

Compliance

Graphics on printed styles conform to OSHA™ standard 29CFR1910.1030.

Specifications

Specifications

OSHA, 29 CFR1910.1030
12 to 14 gal.
30 x 24 in. (76 x 61 cm)
61 cm
24 in.
2 mil
140°C
Elastic
Clear
76 cm
Autoclavable
30 in.
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers