Learn More
Fisherbrand™ Polypropylene, Rectangular Carboy
For Use With: Ideal for storing and dispensing lab water and reagents; Autoclavable; Closure Material: Polypropylene
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 1.17900kg
Description
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
- Durable construction for long service life
- Molded graduations for 1 gallon and 51 increments
- Choice of materials for general use or autoclavability
- Space-saving rectangular design
- Wide mouth closure allows for easy filling and cleaning access
- Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
- Spigot spout without the adapter fits 5/8-in. ID tubing
- Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Specifications
Autoclavable | |
9 L | |
Polypropylene | |
Ideal for storing and dispensing lab water and reagents | |
Wide | |
Rectangular |
2.38 gal. | |
FDA, USP VI | |
White | |
Polypropylene | |
Carboy |