Learn More
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
£12.13 - £125.00
Specifications
Description With Perforated handle
Material Porcelain
Finish Glazed
For Use With (Application) Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric) 1150°C
View More Specs
Products 9
Product Code Brand Height (Metric) Thickness (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Height (Metric) Thickness (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Price Quantity & Availability  
13419709
View Documents
Fisherbrand™
FB965R
5mm 1.75mm 30mm 26mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13439709
View Documents
Fisherbrand™
FB965S
5.5mm 1.75mm 40mm 34mm
£13.75
Pack of 6
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
13429709
View Documents
Fisherbrand™
FB965T
6mm 1.9mm 44mm 38mm
£35.46
Pack of 6
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
13449709
View Documents
Fisherbrand™
FB965U
7mm 2.3mm 53mm 45mm
£35.88
Pack of 6
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
13469709
View Documents
Fisherbrand™
FB965W
7mm 2.5mm 75mm 66mm
£39.05
Pack of 6
Estimated Shipment: 07-08-2025
to see stock.
Add to basket
13459709
View Documents
Fisherbrand™
FB965V
7mm 2.5mm 60mm 54mm
£45.55
Pack of 6
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
13479709
View Documents
Fisherbrand™
FB965X
8mm 2.75mm 87mm 77mm
£125.00
Pack of 6
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
13499709
View Documents
Fisherbrand™
FB965Z
9mm 3mm 115mm 105mm
£23.50
Pack of 6
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
13489709
View Documents
Fisherbrand™
FB965Y
8mm 2.75mm 99mm 90mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
Specifications
With Perforated handle
Glazed
1150°C
Porcelain
Use with Fisherbrand high and wide form porcelain crucible
Crucible Cover
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Porcelain Crucible Covers