Learn More
Fisherbrand™ Clean-Wipes™
Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes.
Brand: Fisherbrand™ 90007-96
15.58 GBP valid until 2024-12-31
Use promo code "24046" to get your promotional price.
Description
Fisherbrand™ Clean-Wipes™ are ideal for cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.
- Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes
- Reclosable container dispenses single wipes, eliminates contamination from dust and dirt
- Premoistened with a blend of pure reagent-grade 70% isopropyl alcohol and 30% deionized water
Order Info
Packed 100 wipes per polyethylene canister
Specifications
Pre-moistened IPA Wipes | |
22.8 cm | |
15.2 cm | |
9 x 6 in. (22.8 x 15.2 cm) | |
For cleaning or wiping optical parts, Glassware, electrodes, cuvettes and microscopes | |
Non-sterile |
9 in. | |
6 in. | |
Polyester Cellulose | |
For cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes. | |
Pure reagent-grade 70% isopropyl alcohol and 30% deionized water |
Safety and Handling
- Premoistened Clean-Wipes
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.