missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Clean-Wipes™
Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes.
£17.46 - £43.77
Specifications
Length (English) | 9 in. |
---|---|
Length (Metric) | 22.8 cm |
Width (English) | 6 in. |
Width (Metric) | 15.2 cm |
Material | Polyester Cellulose |
Product Code | Brand | Product Type | Saturation Solution | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Saturation Solution | Price | Quantity & Availability | |||||
11744346
|
Fisherbrand™
90023-30 |
Dry Wipe | Dry |
£30.58
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
12972780
|
Fisherbrand™
90023-31 |
Pre-moistened DI Wipes | 100% pure deionized water |
£32.92
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11734346
|
Fisherbrand™
2060 |
Pre-moistened DI Wipes | 100% pure deionized water | N/A | ||||||
11754346
|
Fisherbrand™
90007-96 |
Pre-moistened IPA Wipes | Pure reagent-grade 70% isopropyl alcohol and 30% deionized water |
£43.77
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fisherbrand™ Clean-Wipes™ are ideal for cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.
- Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes
- Reclosable container dispenses single wipes, eliminates contamination from dust and dirt
Order Info
Packed 100 wipes per polyethylene canister
Specifications
9 in. | |
6 in. | |
Polyester Cellulose | |
For cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes. | |
Non-sterile |
22.8 cm | |
15.2 cm | |
9 x 6 in. (22.8 x 15.2 cm) | |
For cleaning or wiping optical parts, Glassware, electrodes, cuvettes and microscopes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Clean-Wipes™