Learn More
Fisherbrand™ Polypropylene, Spigot
GREENER_CHOICE
Product Code. 13448039
Spigot; 1/Cs.
Click to view available options
Spigot; 1/Cs.
For Use With: Fisherbrand HDPE and Polypropylene Rectangular Carboys
Value your lab can rely on
Spigots for use with Fisherbrand HDPE and Polypropylene rectangular carboys
  • Spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing
  • Spigot spout without the adapter fits 5/8-in. ID tubing
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With (Equipment) Fisherbrand HDPE and Polypropylene Rectangular Carboys
Material Polypropylene
No. per Case 1 Ea.
Product Type Spigot
Product Content Correction
Product Title
Fisherbrand™ Polypropylene, Spigot >