Description
Value your lab can rely on
Closures for use with Fisherbrand HDPE and Polypropylene rectangular carboys
- Closure for the Polypropylene carboys for use when autoclaving with liquid contained inside
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | Autoclavable |
For Use With (Equipment) | Fisherbrand HDPE and Polypropylene Rectangular Carboys |
Material | Polypropylene |
Product Type | Closure |