missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Reusable Amber Glass Media Bottles
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Description
Fisherbrand™ Reusable Amber Glass Media Bottles with cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-free, replaceable polypropylene pour ring.
- Permanent white enamel marking spot
- GL 45 screw thread
- Linerless
- Autoclavable (to 140°C)
- Amber borosilicate glass
- Polypropylene cap
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Amber |
| For Use With (Application) | Media and reagent storage, Holding and dispensing fluids, Chemical storage |
| Includes | Screw Cap, Pouring Ring |
| Material | Borosilicate Glass (3.3) |
| No. per Case | 10/Cs. |
| Screw Color | Blue |
| Screw Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Color | Blue |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction