Learn More
Fisherbrand™ Rotary Mixer Discs
Suitable for end-to-end mixing tubes
Brand:  Fisherbrand™ RMD012
 View more versions of this product
Product Code. 12387488
  • £115.00 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rotary Mixer Disc complete with clips for tubes, supplied unassembled
  • Suitable for 'End to End Mixing' tubes,
  • Clips for tube diameters between  8 - 11 mm are suitable for 0.5mL, 1.5mL and 2.0mL Microcentrifuge tubes, 3mL Vacutainers and 5mL capped tubes (8 -11 mm)
  • Clips for tube diameters between 15 - 17 mm are suitable for 15 mL centrifuge tubes
  • Easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
Specifications
Rotary Mixer Disc
Rotary Mixer Disc
Tubes with a Diameter Between 15-17 mm