Learn More
Fisherbrand™ Round Cuvets
For use with commercial colorimeters and spectrophotometers.
£382.00
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
16333416
|
Fisherbrand™
|
19 mm | 105 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
For use with commercial colorimeters and spectrophotometers.
- Reference mark near top assists repositioning
- Manufactured from borosilicate glass
- Available in case of 36
Specifications
Cuvette | |
Borosilicate Glass | |
Round |