missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rubber-Lined Plastic Caps

Product Code. 11588212
Click to view available options
Neck Finish:
22-400
38-400
58-400
This item is not returnable. View return policy
This item is not returnable. View return policy

Resilient, resistant to moisture vapor

  • Black, rubber-lined caps Autoclavable
  • Packaged in 12 packs

Specifications

Color Black
Description Screw cap
Material Plastic
No. per Case 576 Cs.
Quantity 576/PK.
Sterility Sterile
Liner Material Rubber
Neck Finish 38-400
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Bottle
Closure Liner 14B Rubber
Closure Material Plastic
Closure Size 38-400 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'