Learn More
Fisherbrand™ Safety Bulb Pipette Filler
Fill any standard size pipet without the risk of cracking or chipping. Fisherbrand™ Safety Bulb Pipet Filler uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Comes with an elastic band for convenient hanging on bottles or wrist.
Brand: Fisherbrand™ HS20623
22.08 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Includes an elastic band for convenient hanging on bottles or wrist. Allows you to fill any standard size pipet without the risk of cracking or chipping.
- Pipette does not contact hard plastic, which reduces the risk of cracking and chipping
- 15 mL capacity, 106 mm L x 41 mm OD (4.5 to 6.5 mm hole dia.)
- Elastic band for hanging on reagent bottle or wrist
- Filler fits standard size pipettes
Specifications
4.5 to 6.5 mm | |
Pipette | |
Bulb | |
Pipette Filler | |
Standard Size Pipettes | |
3 Pk. | |
107 mm | |
1.6 in. | |
41 mm |
Red | |
Natural Rubber, ABS | |
1 | |
15 mL | |
Handy elastic band for hanging on wrist or reagent bottle | |
4.21 in. | |
0.2 to 0.3 in. | |
4.5 to 6.5 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.