missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles

Product Code. 12349185
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Color:
Assorted
Blue
Green
Natural
Red
Yellow
For Use With (Application):
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
Variety
This item is not returnable. View return policy
This item is not returnable. View return policy

Keep liquids in clearly marked bottles that help prevent accidental misuse. Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and Label Your Own bottles.

Keep liquids in clearly marked bottles that help prevent accidental misuse. Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and Label Your Own bottles.

 

  • Feature a wide mouth 53mm (2 in.) for easy refilling
  • Bottles are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Color-coded polypropylene closures aid in quick and easy identification
  • Note: Use LYOB only with solvents compatible with LDPE

 

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity 16 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Yellow, Natural
For Use With (Application) Isopropanol
Material LDPE
Quantity 6/Pk.
Closure Color Yellow
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing Yes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'