missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Color:
Assorted
Blue
Green
Natural
Red
Yellow
For Use With (Application):
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
Variety
Description
Keep liquids in clearly marked bottles that help prevent accidental misuse. Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.
- Feature a wide mouth 53mm (2 in.) for easy refilling
- Bottles are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Color-coded polypropylene closures aid in quick and easy identification
- Note: Use LYOB only with solvents compatible with LDPE
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Capacity | 32 oz., 1000mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Green, Natural |
For Use With (Application) | Methanol |
Material | LDPE |
Quantity | 4/Pk. |
Closure Color | Green |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Closure Included | Yes |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'