Learn More
Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles
Eliminates drips and allows bottle to dispense upright or inverted. Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.
£37.71 - £37.79
Specifications
Capacity | 16 oz., 500mL |
---|---|
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Material | LDPE |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Produktcode | Marke | Capacity (Metric) | Chemical Label | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Capacity (Metric) | Chemical Label | Preis | Menge & Verfügbarkeit | ||||||
12369195
|
Fisherbrand™
11816-3019 |
500 mL | Ethanol | N/A | N/A | N/A | |||||
12309205
|
Fisherbrand™
11816-3004 |
500 mL | Distilled Water | N/A | N/A | N/A | |||||
12399195
|
Fisherbrand™
11816-3015 |
500 mL | Sodium Hypochlorite | N/A | N/A | N/A | |||||
12379195
|
Fisherbrand™
11816-3008 |
500 mL | Isopropanol | N/A | N/A | N/A | |||||
12359195
|
Fisherbrand™
11816-3050 |
500 mL | Assorted | N/A | N/A | N/A | |||||
12319205
|
Fisherbrand™
11816-3001 |
500 mL | Acetone | N/A | N/A | N/A | |||||
Beschreibung
Eliminates drips and allows bottle to dispense upright or inverted. Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.
- Valve allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent LDPE bottles with PP valves; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Spezifikation
16 oz., 500mL | |
16 oz. | |
OSHA 29 CFR 1910.1200 | |
Not Autoclavable | |
Wide | |
Wash Bottle | |
53 mm |
500 mL | |
LDPE | |
Yes | |
No | |
Cylindrical | |
Polypropylene | |
Yes |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.