missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Silicone Recessed Septum Stoppers

Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal

Brand:  Fisherbrand™ FB57897

 View more versions of this product

Product Code. 11562503

  • £50.50 / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Elliptical flange to allow efficient removal of stopper from aperature
  • Easily pierced with a hypodermic needle

Specifications

Specifications

14.50 to 15.50 mm
For 14.5 to 15.5mm I.D. containers
Translucent
Septum Stopper
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
1
Silicone
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.