missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Small Round Biohazard Labels
Shop All Fisherbrand Products

Click to view available options
Color:
Black, Red
Unit Size:
Pack of 1000
Description
- Strong adhesive ensures that labels remain in place
- Red labels with black 1 in. core
- Round, small, 1 in. dia.
Compliance
Meet OSHA requirements.
Specifications
Specifications
| Certifications/Compliance | OSHA |
| Color | Black, Red |
| Format | Label |
| Material | Paper |
| Product Type | Round Biohazard Label |
| For Use With (Application) | Biohazard |
| Quantity | 1000/Roll |
| Shape | Round |
| Diameter (English) | 1.25 in. |
| Diameter (Metric) | 3.1 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction