missing translation for 'onlineSavingsMsg'
Learn More

LevGo™ Inoculation Picks

Isolate individual colonies with picks manufactured from White Birch wood. LevGo™ Inoculation Picks are naturally biodegradable and disposable, but are also autoclavable and suitable for food contact.

Brand:  LevGo™ HS120012

 View more versions of this product

Product Code. 15316568

  • £186.00 / Pack of 1500
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Available in three lengths and two needle sizes to suit a wide range of application needs.
  • Naturally biodegradable, made from 100% U.S.A grown white birch wood
  • Manufactured in an FDA-registered factory and comply with FDA standards for food contact
  • Available in semi or needle point styles for delicate operations
  • Autoclavable
  • Disposable
Specifications

Specifications

Wooden Pick
Standard
1500 Pk.
63.5 mm
White Birch Wood
Needle point
2.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.