missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
Click to view available options
For Use With (Application):
Acetone
Ethanol
Isopropanol
Methanol
Unlabeled
Description
Are pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
- Wide mouth 2 in. (53mm) bottle for easy refilling
- Polypropylene closure with leakproof design, color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Order Info
Prelabeled with six common solvents; also available unlabeled.
Specifications
Capacity | 16 oz., 500mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
For Use With (Application) | Ethanol |
Material | LDPE |
Closure Color | Natural |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'